missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-5-fluorobenzonitrile, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
2-Bromo-5-fluorobenzonitrile is used in the synthesis of pyrimidine functional compounds acting as GABAAα2/α3 binding site agonists used in the treatment of anxiety disorders.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Bromo-5-fluorobenzonitrile is used in the synthesis of pyrimidine functional compounds acting as GABAAα2/α3 binding site agonists used in the treatment of anxiety disorders.
Solubility
Soluble in methanol.
Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
2-Bromo-5-fluorobenzonitrile is used in the synthesis of pyrimidine functional compounds acting as GABAAα2/α3 binding site agonists used in the treatment of anxiety disorders.
Solubility
Soluble in methanol.
Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Specifications
Melting Point | 88°C to 92°C |
Quantity | 5 g |
UN Number | UN3439 |
Beilstein | 8678330 |
Solubility Information | Soluble in methanol. |
Formula Weight | 200.01 |
Percent Purity | ≥98% |
Chemical Name or Material | 2-Bromo-5-fluorobenzonitrile |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'