missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-6-methoxynaphthalene, 98%, Thermo Scientific Chemicals

Catalog Number 10021711
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
5111-65-9
MFCD00004062
AYFJBMBVXWNYLT-UHFFFAOYSA-N
6-methoxy-2-bromonaphthalene, naphthalene, 2-bromo-6-methoxy, 6-bromo-2-methoxynaphthalene, 2-bromo-6-methoxy-naphthalene, 6-brom-2-naphthyl-methyl ether, 2-bromo-6-methoxynapthalene, unii-s19t080mrg, naproxen ep impurity n, bromo 2--6-methoxy naphthalene, 2-methoxy-6-bromonaphthalene
78786
2-bromo-6-methoxynaphthalene
COC1=CC2=C(C=C1)C=C(C=C2)Br
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
5111-65-9
MFCD00004062
AYFJBMBVXWNYLT-UHFFFAOYSA-N
6-methoxy-2-bromonaphthalene, naphthalene, 2-bromo-6-methoxy, 6-bromo-2-methoxynaphthalene, 2-bromo-6-methoxy-naphthalene, 6-brom-2-naphthyl-methyl ether, 2-bromo-6-methoxynapthalene, unii-s19t080mrg, naproxen ep impurity n, bromo 2--6-methoxy naphthalene, 2-methoxy-6-bromonaphthalene
78786
2-bromo-6-methoxynaphthalene
COC1=CC2=C(C=C1)C=C(C=C2)Br

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 105°C to 111°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C11H9BrO
Linear Formula BrC10H6OCH3
Quantity 5 g
Beilstein 06, 651
Molecular Weight (g/mol) 237.1
Formula Weight 237.1
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-Bromo-6-methoxynaphthalene, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Bromo-6-methoxynaphthalene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'