missing translation for 'onlineSavingsMsg'
Learn More

2-Bromoanthracene, 98%, Thermo Scientific Chemicals

Catalog Number p-7077517
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
7321-27-9
C14H9Br
257.13
MFCD07784002
PYXBCVWIECUMDW-UHFFFAOYSA-N
2-bromo-anthracene, anthracene, 2-bromo, pubchem19987, acmc-209opw, ksc377a5l
12346099
2-bromoanthracene
BrC1=CC2=CC3=CC=CC=C3C=C2C=C1
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
7321-27-9
C14H9Br
257.13
MFCD07784002
PYXBCVWIECUMDW-UHFFFAOYSA-N
2-bromo-anthracene, anthracene, 2-bromo, pubchem19987, acmc-209opw, ksc377a5l
12346099
2-bromoanthracene
BrC1=CC2=CC3=CC=CC=C3C=C2C=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 220.0°C to 221.5°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 257.13
Percent Purity 98%
Chemical Name or Material 2-Bromoanthracene
compliance-icons
Product Identifier
  • 2-Bromoanthracene, 98%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'