missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ (2-Bromoethoxy)-tert-butyldimethylsilane, 98%, stab. with sodium carbonate

Brand:  Alfa Aesar™ H56594.03

Code : NEW

Additional Details : CAS Number : 86864-60-0 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15472786

  • 29.00 € / 1g


Estimated Shipment date
from Supplier 22-05-2019
Add to basket

Specifications

Chemical Identifiers

86864-60-0
239.228
JBKINHFZTVLNEM-UHFFFAOYSA-N
3608067
CC(C)(C)[Si](C)(C)OCCBr
C8H19BrOSi
MFCD00209550
2-bromoethoxy-tert-butyldimethylsilane, 2-bromoethoxy tert-butyl dimethylsilane, 2-bromoethoxy-t-butyl dimethylsilane, 2-t-butyldimethylsiloxy ethylbromide, 2-bromoethoxy-tert-butyldimethylsilane, 2-bromoethoxy-t-butyldimethylsilane, 2-bromoethoxy-t-butyldimethylsilane, 2-bromo-ethoxy-tert-butyl-dimethyl-silane, silane, 2-bromoethoxy 1,1-dimethylethyl dimethyl, brch2ch2otbs
2-bromoethoxy-tert-butyl-dimethylsilane

Specifications

(2-Bromoethoxy)-tert-butyldimethylsilane
86864-60-0
MFCD00209550
JBKINHFZTVLNEM-UHFFFAOYSA-N
2-bromoethoxy-tert-butyl-dimethylsilane
3608067
Liquid
1.115g/mL
120°C (100mmHg)
1g
Stabilized with sodium carbonate
C8H19BrOSi
2-bromoethoxy-tert-butyldimethylsilane, 2-bromoethoxy tert-butyl dimethylsilane, 2-bromoethoxy-t-butyl dimethylsilane, 2-t-butyldimethylsiloxy ethylbromide, 2-bromoethoxy-tert-butyldimethylsilane, 2-bromoethoxy-t-butyldimethylsilane, 2-bromoethoxy-t-butyldimethylsilane, 2-bromo-ethoxy-tert-butyl-dimethyl-silane, silane, 2-bromoethoxy 1,1-dimethylethyl dimethyl, brch2ch2otbs
CC(C)(C)[Si](C)(C)OCCBr
239.228
239.23
98%
1.444
Colorless
Videos
SDS