missing translation for 'onlineSavingsMsg'
Learn More

2-Bromomethyl-1,3-dioxolane, 97%, Thermo Scientific Chemicals

Catalog Number p-75597
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
4360-63-8
C4H7BrO2
167
MFCD00003214
CKIIJIDEWWXQEA-UHFFFAOYSA-N
78068
2-(bromomethyl)-1,3-dioxolane
C1COC(O1)CBr
This item is not returnable. View return policy
4360-63-8
C4H7BrO2
167
MFCD00003214
CKIIJIDEWWXQEA-UHFFFAOYSA-N
78068
2-(bromomethyl)-1,3-dioxolane
C1COC(O1)CBr

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Color Colorless
Density 1.6100g/mL
Boiling Point 80°C to 82°C (27.0 mmHg)
Flash Point 62°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4810 to 1.483
Quantity 25 mL
Beilstein 19, II, 8
Specific Gravity 1.61
Synonym 2-bromomethyl-1,3-dioxolane, bromoacetaldehyde ethylene acetal, 1,3-dioxolane, 2-bromomethyl, 1,3-dioxolan-2-yl methyl bromide, bromomethyl dioxolane, acmc-1afrf, 2-bromomethyl-1,3-dioxolan, ksc495q7b, 2-bromomethyl-1,3 dioxolane, 2-bromomethyl-1, 3-dioxolane
Formula Weight 167
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Bromomethyl-1, 3-dioxolane, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Bromomethyl-1,3-dioxolane
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'