Learn More
2-(Bromomethyl)-1-methyl-1H-benzimidazole hydrobromide, 97%, Maybridge™
Brand: Maybridge CC03858CB
Code : 31
Additional Details : CAS Number : 934570-40-8 Weight : 0.05025kg Transport : UN number : 2811 Chem class : 6.1 Pack group : III
Packaging | Amber Glass Bottle |
---|---|
Quantity | 250mg |
Chemical Identifiers
934570-40-8 | |
306.001 | |
WHMCWOHHFOGLGP-UHFFFAOYSA-N | |
24229451 | |
CN1C2=CC=CC=C2N=C1CBr.Br |
C9H10Br2N2 | |
MFCD09702343 | |
2-bromomethyl-1-methyl-1h-benzimidazole hydrobromide, 2-bromomethyl-1-methyl-1h-benzo d imidazole hydrobromide, 2-bromomethyl-1-methyl-1,3-benzodiazole hydrobromide, 2-bromomethyl-1-methyl-benzimidazole hydrobromide, 2-bromomethyl-1-methyl-1h-benzimidazole hbr, 2-bromomethyl-1-methyl-1h-benzimidazole-hydrogen bromide 1/1 | |
2-(bromomethyl)-1-methylbenzimidazole;hydrobromide |
Specifications
2-(Bromomethyl)-1-methyl-1H-benzimidazole hydrobromide | |
250mg | |
96.5 | |
97% | |
MFCD09702343 | |
WHMCWOHHFOGLGP-UHFFFAOYSA-N | |
2-(bromomethyl)-1-methylbenzimidazole;hydrobromide | |
24229451 | |
97% |
Amber Glass Bottle | |
934570-40-8 | |
100.0 | |
C9H10Br2N2 | |
2-bromomethyl-1-methyl-1h-benzimidazole hydrobromide, 2-bromomethyl-1-methyl-1h-benzo d imidazole hydrobromide, 2-bromomethyl-1-methyl-1,3-benzodiazole hydrobromide, 2-bromomethyl-1-methyl-benzimidazole hydrobromide, 2-bromomethyl-1-methyl-1h-benzimidazole hbr, 2-bromomethyl-1-methyl-1h-benzimidazole-hydrogen bromide 1/1 | |
CN1C2=CC=CC=C2N=C1CBr.Br | |
306.001 | |
306 |