missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromophenylacetic acid, 98+%, Thermo Scientific Chemicals
25.53 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
100 g
Description
Employed as a pharmaceutical intermediate. It is also used in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Employed as a pharmaceutical intermediate. It is also used in organic synthesis.
Solubility
Very faint turbidity in methanol.
Notes
Store away from oxidizing agents. Keep the container tightly sealed and store in cool, dry condition in well ventilated place.
Employed as a pharmaceutical intermediate. It is also used in organic synthesis.
Solubility
Very faint turbidity in methanol.
Notes
Store away from oxidizing agents. Keep the container tightly sealed and store in cool, dry condition in well ventilated place.
Specifications
Melting Point | 105°C to 106°C |
Quantity | 5 g |
Beilstein | 2250655 |
Solubility Information | Very faint turbidity in methanol. |
Formula Weight | 215.05 |
Percent Purity | ≥98% |
Chemical Name or Material | 2-Bromophenylacetic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'