missing translation for 'onlineSavingsMsg'
Learn More

2-Butanol, Reagent Grade, ≥99%, Honeywell™

Catalog Number p-7120055
Click to view available options
Quantity:
1 L
2.5 L
6 x 1 L
Packaging:
Glass Bottle
This item is not returnable. View return policy
78-92-2
C4H10O
74.12
MFCD00004569
BTANRVKWQNVYAZ-UHFFFAOYNA-N
2-butanol, sec-butanol, sec-butyl alcohol, 2-hydroxybutane, 2-butyl alcohol, s-butyl alcohol, butylene hydrate, s-butanol, 1-methyl-1-propanol, 1-methylpropyl alcohol
6568
CHEBI:35687
butan-2-ol
CCC(C)O
This item is not returnable. View return policy
78-92-2
C4H10O
74.12
MFCD00004569
BTANRVKWQNVYAZ-UHFFFAOYNA-N
2-butanol, sec-butanol, sec-butyl alcohol, 2-hydroxybutane, 2-butyl alcohol, s-butyl alcohol, butylene hydrate, s-butanol, 1-methyl-1-propanol, 1-methylpropyl alcohol
6568
CHEBI:35687
butan-2-ol
CCC(C)O

Reagent Grade,≥99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY
pH neutral
Melting Point -115°C
Color Colorless
Linear Formula CH3CH2CH(OH)CH3
UN Number UN1120
Beilstein 773649
Formula Weight 74.12g/mol
Percent Purity ≥99%
Density 0.808g/mL at 25°C
Boiling Point 98°C
Quantity 1 L
Explosion Limit 0.098
Flash Point 27°C (80.6°F)
Ignition Point 761°F
Odor Characteristic
Packaging Glass Bottle
Physical Form Liquid
Vapor Pressure 12.5mmHg (20°C)
Chemical Name or Material 2-Butanol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Butanol
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'