missing translation for 'onlineSavingsMsg'
Learn More

cis-2-Butene-1,4-diol, 97%, ACROS Organics™

Brand:  Acros Organics 107720025

Code : 29

Additional Details : CAS Number : 6117-80-2 Weight : 2.72500kg


Packaging Glass bottle
Quantity 2.5L

 View more versions of this product

Product Code. 11954631

  • 168.00 € / 2.50 L

Estimated Shipment: 06-07-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

6117-80-2
88.11
ORTVZLZNOYNASJ-UPHRSURJSA-N
643790
OC\C=C/CO
C4H8O2
MFCD00002924,MFCD00063207
cis-2-butene-1,4-diol, z-2-butene-1,4-diol, cis-butenediol, unii-za7vgu6scv, cis-1,4-dihydroxy-2-butene, z-but-2-ene-1,4-diol, 2-butene-1,4-diol, 2z-but-2-ene-1,4-diol, 2-butene-1,4-diol, z, za7vgu6scv
(Z)-but-2-ene-1,4-diol

Specifications

cis-2-Butene-1, 4-diol
128°C
Glass bottle
1.07
235.0°C
7.0°C
97% cis
96% min. (cis-isomer) (GC)
HOCH2CH=CHCH2OH
cis-2-butene-1,4-diol, z-2-butene-1,4-diol, cis-butenediol, unii-za7vgu6scv, cis-1,4-dihydroxy-2-butene, z-but-2-ene-1,4-diol, 2-butene-1,4-diol, 2z-but-2-ene-1,4-diol, 2-butene-1,4-diol, z, za7vgu6scv
ORTVZLZNOYNASJ-UPHRSURJSA-N
(Z)-but-2-ene-1,4-diol
643790
Liquid
1.0700g/mL
Authentic
1.4770 to 1.4800
0.3% max.
Colorless
2.5L
6117-80-2
C4H8O2
MFCD00002924,MFCD00063207
Solubility in water: soluble. Other solubilities: soluble in alcohol and acetone,slightly soluble in benzene
OC\C=C/CO
88.11
88.11
97%
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • cis-2-Butene-1,4-diol

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

SDS Documents
Certificates