missing translation for 'onlineSavingsMsg'
Learn More

(2-Butyl-1H-imidazol-4-yl)methanol, 97%, Maybridge™

Brand:  Maybridge SB02033DA

Code : 31

Additional Details : CAS Number : 68283-19-2 Weight : 0.05100kg

Quantity 1g

 View more versions of this product

Product Code. 10029481

  • 13.91 € / 1g

Estimated Shipment
20-05-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

68283-19-2
154.213
UZKBZGAMRJRWLR-UHFFFAOYSA-N
2735673
CCCCC1=NC=C(N1)CO
C8H14N2O
MFCD00239516
2-butyl-1h-imidazol-4-yl methanol, 2-butyl-5-hydroxymethylimidazole, 2-butyl-1h-imidazol-5-yl methanol, unii-k14mm5ob9a, 2-butyl-4-hydroxymethyl imidazole, k14mm5ob9a, 2-butylimidazol-5-yl methan-1-ol, 2-butyl-3h-imidazol-4-yl methanol, 2-n-butyl-4-imidazolemethanol, 2-n-butyl-4-hydroxymethylimidazole
(2-butyl-1H-imidazol-5-yl)methanol

Specifications

(2-Butyl-1H-imidazol-4-yl)methanol
1g
97.0
C8H14N2O
2-butyl-1h-imidazol-4-yl methanol, 2-butyl-5-hydroxymethylimidazole, 2-butyl-1h-imidazol-5-yl methanol, unii-k14mm5ob9a, 2-butyl-4-hydroxymethyl imidazole, k14mm5ob9a, 2-butylimidazol-5-yl methan-1-ol, 2-butyl-3h-imidazol-4-yl methanol, 2-n-butyl-4-imidazolemethanol, 2-n-butyl-4-hydroxymethylimidazole
CCCCC1=NC=C(N1)CO
154.213
154.21
Amber Glass Bottle
68283-19-2
97%
MFCD00239516
UZKBZGAMRJRWLR-UHFFFAOYSA-N
(2-butyl-1H-imidazol-5-yl)methanol
2735673
97%
Videos
SDS Documents