missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-2-methylpropane, 99%, Thermo Scientific Chemicals

Catalog Number p-88477
Click to view available options
Quantity:
25 mL
500 mL
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
507-20-0
C4H9Cl
92.57
MFCD00000816
NBRKLOOSMBRFMH-UHFFFAOYSA-N
tert-butyl chloride, propane, 2-chloro-2-methyl, tert-butylchloride, chlorotrimethylmethane, trimethylchloromethane, 2-chloroisobutane, t-butyl chloride, t-butylchloride, 2-methyl-2-chloropropane, 2-methyl-2-propyl chloride
10486
2-chloro-2-methylpropane
CC(C)(C)Cl
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
507-20-0
C4H9Cl
92.57
MFCD00000816
NBRKLOOSMBRFMH-UHFFFAOYSA-N
tert-butyl chloride, propane, 2-chloro-2-methyl, tert-butylchloride, chlorotrimethylmethane, trimethylchloromethane, 2-chloroisobutane, t-butyl chloride, t-butylchloride, 2-methyl-2-chloropropane, 2-methyl-2-propyl chloride
10486
2-chloro-2-methylpropane
CC(C)(C)Cl

2-Chloro-2-methylpropane, 99%, C4H9Cl, CAS Number-507-20-0, 2-methyl-2-chloropropane, propane, 2-chloro-2-methyl, 2-chloroisobutane, tert-butylchloride, t-butylchloride, chlorotrimethylmethane, trimethylchloromethane, tert-butyl chloride, t-butyl chloride, 2-methyl-2-propyl chloride, 500mL

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point -25.0°C
Density 0.8500g/mL
Boiling Point 51.0°C to 52.0°C
Flash Point −27°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)3CCl
Refractive Index 1.3838 to 1.3858
Quantity 25 mL
Beilstein 01, 125
Fieser 02,46
Merck Index 15, 1563
Specific Gravity 0.85
Solubility Information Solubility in water: slightly soluble. Other solubilities: miscible with alcohol and ether
Viscosity 0.51 mPa.s (20°C)
Formula Weight 92.57
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Chloro-2-methylpropane, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Chloro-2-methylpropane
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Supplemental information
  • MIXTURE LIST-Contains : terrt-butyl chloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'