missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3,6-difluorobenzonitrile, 97%, Thermo Scientific™

Catalog Number p-7029142
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
886501-33-3
C7H2ClF2N
173.55
MFCD06660175
AREWFYUZFHLWNH-UHFFFAOYSA-N
2-chloro-3,6-difluoro-benzonitrile, 2-chloro-3,6-difluorobenzenecarbonitrile, 2-chloro-3,6-difluorobenzonitrile
17750746
2-chloro-3,6-difluorobenzonitrile
FC1=C(Cl)C(C#N)=C(F)C=C1
This item is not returnable. View return policy
886501-33-3
C7H2ClF2N
173.55
MFCD06660175
AREWFYUZFHLWNH-UHFFFAOYSA-N
2-chloro-3,6-difluoro-benzonitrile, 2-chloro-3,6-difluorobenzenecarbonitrile, 2-chloro-3,6-difluorobenzonitrile
17750746
2-chloro-3,6-difluorobenzonitrile
FC1=C(Cl)C(C#N)=C(F)C=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
UN Number UN3276
Formula Weight 173.55
Percent Purity 97%
Quantity 1 g
Chemical Name or Material 2-Chloro-3,6-difluorobenzonitrile
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'