missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3-fluorobenzonitrile, 97%, Thermo Scientific™

Catalog Number 15475927
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
874781-08-5
C7H3ClFN
155.56
MFCD03407966
ROXFZJBRIXTCPI-UHFFFAOYSA-N
3-fluoro-2-chlorobenzonitrile, 2-chloro-3-fluoro-benzonitrile, buttpark 45\01-05, benzonitrile, 2-chloro-3-fluoro, pubchem3554, benzonitrile,2-chloro-3-fluoro, 2-chloro-3-fluorobenzenecarbonitrile
3723586
2-chloro-3-fluorobenzonitrile
FC1=CC=CC(C#N)=C1Cl
This item is not returnable. View return policy
874781-08-5
C7H3ClFN
155.56
MFCD03407966
ROXFZJBRIXTCPI-UHFFFAOYSA-N
3-fluoro-2-chlorobenzonitrile, 2-chloro-3-fluoro-benzonitrile, buttpark 45\01-05, benzonitrile, 2-chloro-3-fluoro, pubchem3554, benzonitrile,2-chloro-3-fluoro, 2-chloro-3-fluorobenzenecarbonitrile
3723586
2-chloro-3-fluorobenzonitrile
FC1=CC=CC(C#N)=C1Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 55°C to 56°C
Color White
UN Number UN3439
Formula Weight 155.56
Percent Purity 97%
Quantity 250 mg
Physical Form Powder
Chemical Name or Material 2-Chloro-3-fluorobenzonitrile
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'