missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3-methylbenzotrifluoride, 95%, Acros Organics

Brand:  Acros Organics 451290050

Code : NEW

Additional Details : CAS Number : 74483-48-0 Weight : 0.01000kg


Quantity 5g

 View more versions of this product

Product Code. 15269696

  • 370.80 € / 5g

Estimated Shipment
22-07-2020

Please to purchase this item. Need a web account? Register with us today!



Specifications

Chemical Identifiers

74483-48-0
194.58
2-chloro-3-methylbenzotrifluoride, 2-chloro-1-methyl-3-trifluoromethyl benzene, chloro-3-methyl benzotrifluoride
2-chloro-1-methyl-3-(trifluoromethyl)benzene
C8H6ClF3
NJLAMXMEANIPBX-UHFFFAOYSA-N
12712336
CC1=C(C(=CC=C1)C(F)(F)F)Cl

Specifications

2-Chloro-3-methylbenzotrifluoride
82°C to 84°C (40.0 mBar)
74483-48-0
C8H6ClF3
NJLAMXMEANIPBX-UHFFFAOYSA-N
2-chloro-1-methyl-3-(trifluoromethyl)benzene
12712336
95%
64°C
5g
94% min. (GC)
2-chloro-3-methylbenzotrifluoride, 2-chloro-1-methyl-3-trifluoromethyl benzene, chloro-3-methyl benzotrifluoride
CC1=C(C(=CC=C1)C(F)(F)F)Cl
194.58
194.58
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 2-Chloro-3-methylbenzotrifluoride, 95%

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

Documents