missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-6-fluoro-3-methoxybenzaldehyde, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
Descripción
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
2-Chloro-6-fluorobenzaldehyde was used in the synthesis of novel copolymers of methyl 2-cyano-3-dihalophenyl-2-propenoates and styrene.
Solubility
Soluble in water 15 mg/L at 20°C . Also soluble in heptane, toluene, methanol, ethyl acetate, acetone.
Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases.
2-Chloro-6-fluorobenzaldehyde was used in the synthesis of novel copolymers of methyl 2-cyano-3-dihalophenyl-2-propenoates and styrene.
Solubility
Soluble in water 15 mg/L at 20°C . Also soluble in heptane, toluene, methanol, ethyl acetate, acetone.
Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases.
Especificaciones
Melting Point | 104°C to 107°C |
Quantity | 1 g |
Sensitivity | Air Sensitive |
Solubility Information | Soluble in water 15mg/L at 20°C . Also soluble in heptane,toluene,methanol,ethyl acetate,acetone. |
Formula Weight | 188.59 |
Percent Purity | 97% |
Chemical Name or Material | 2-Chloro-6-fluoro-3-methoxybenzaldehyde |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'