missing translation for 'onlineSavingsMsg'
Learn More

2-chloro-6-fluorobenzaldehyde, 95%, Thermo Scientific Chemicals

Catalog Number p-3777880
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
387-45-1
C7H4ClFO
158.56
MFCD00003306
OACPOWYLLGHGCR-UHFFFAOYSA-N
benzaldehyde, 2-chloro-6-fluoro, 2,6-cfad, 2-chloro-6-fluoro-benzaldehyd, unii-51yj9bw8w7, 2-chloro-6-fluoro benzaldehyde, 2-chloro-6-fluoro-benzaldehyde, 2-fluoro-6-chloro benzaldehyde, 6-chloro-2-fluorobenzaldehyde, pubchem1434, acmc-209j1r
67847
2-chloro-6-fluorobenzaldehyde
FC1=CC=CC(Cl)=C1C=O
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
387-45-1
C7H4ClFO
158.56
MFCD00003306
OACPOWYLLGHGCR-UHFFFAOYSA-N
benzaldehyde, 2-chloro-6-fluoro, 2,6-cfad, 2-chloro-6-fluoro-benzaldehyd, unii-51yj9bw8w7, 2-chloro-6-fluoro benzaldehyde, 2-chloro-6-fluoro-benzaldehyde, 2-fluoro-6-chloro benzaldehyde, 6-chloro-2-fluorobenzaldehyde, pubchem1434, acmc-209j1r
67847
2-chloro-6-fluorobenzaldehyde
FC1=CC=CC(Cl)=C1C=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 94.0
CAS Max % 100.0
Melting Point 34°C to 39°C
Color White to Yellow
Boiling Point 92°C (10.0 mmHg)
Flash Point 101°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula ClC6H3(F)CHO
Quantity 25 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in acetone,ether,chloroform
Formula Weight 158.56
Percent Purity 95%
Physical Form Crystalline Solid
Chemical Name or Material 2-Chloro-6-fluorobenzaldehyde
Show More Show Less
compliance-icons
Product Identifier
  • 2-Chloro-6-fluorobenzaldehyde
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'