missing translation for 'onlineSavingsMsg'
Learn More

2-chloro-6-fluorobenzyl bromide, 96%, Thermo Scientific™

Catalog Number p-4323247
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
68220-26-8
C7H5BrClF
223.47
MFCD00040126
IGUVNNXFTDCASP-UHFFFAOYSA-N
2-chloro-6-fluorobenzyl bromide, 2-bromomethyl-1-chloro-3-fluorobenzene, 2-chloro-6-fluorobenzylbromide, 2-bromomethyl-1-chloro-3-fluoro-benzene, benzene, 2-bromomethyl-1-chloro-3-fluoro, 2-bromomethyl-3-chloro-1-fluorobenzene, alpha-bromo-2-chloro-6-fluorotoluene, pubchem4894, acmc-1bdpq
2773625
2-(bromomethyl)-1-chloro-3-fluorobenzene
FC1=CC=CC(Cl)=C1CBr
This item is not returnable. View return policy
68220-26-8
C7H5BrClF
223.47
MFCD00040126
IGUVNNXFTDCASP-UHFFFAOYSA-N
2-chloro-6-fluorobenzyl bromide, 2-bromomethyl-1-chloro-3-fluorobenzene, 2-chloro-6-fluorobenzylbromide, 2-bromomethyl-1-chloro-3-fluoro-benzene, benzene, 2-bromomethyl-1-chloro-3-fluoro, 2-bromomethyl-3-chloro-1-fluorobenzene, alpha-bromo-2-chloro-6-fluorotoluene, pubchem4894, acmc-1bdpq
2773625
2-(bromomethyl)-1-chloro-3-fluorobenzene
FC1=CC=CC(Cl)=C1CBr
CAS Min % 95.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.6290g/mL
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Linear Formula Cl(F)C6H3CH2Br
Refractive Index 1.569
Quantity 1 g
Specific Gravity 1.629
Formula Weight 223.47
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 2-chloro-6-fluorobenzyl bromide
Mostrar más Mostrar menos
compliance-icons
Nombre del producto
  • 2-Chloro-6-fluorobenzyl bromide
Palabra de advertencia
  • Danger
Clasificación
  • Skin corrosion/irritation Category 1 B
Indicaciones de peligro
  • H314-Causes severe skin burns and eye damage.
Consejos de prudencia
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'