missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Chloro-N-(3,4-difluorophenyl)benzamide, 97%

Brand:  Alfa Aesar™ H59688.03

Code : NEW

Additional Details : CAS Number : 330467-98-6 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15416078

  • 448.12 € / 1g


Estimated Shipment date
from Supplier 21-11-2018
Add to basket

Specifications

Chemical Identifiers

330467-98-6
267.66
HFCQDEOKINDBAS-UHFFFAOYSA-N
4223591
C1=CC=C(C(=C1)C(=O)NC2=CC(=C(C=C2)F)F)Cl
C13H8ClF2NO
MFCD01213292
2-chloro-n-3,4-difluorophenyl benzamide, 2-chloro-n~1~-3,4-difluorophenyl benzamide, n-3,4-difluorophenyl 2-chlorophenyl carboxamide
2-chloro-N-(3,4-difluorophenyl)benzamide

Specifications

2-Chloro-N-(3,4-difluorophenyl)benzamide
330467-98-6
MFCD01213292
HFCQDEOKINDBAS-UHFFFAOYSA-N
2-chloro-N-(3,4-difluorophenyl)benzamide
4223591
Solid
1g
C13H8ClF2NO
2-chloro-n-3,4-difluorophenyl benzamide, 2-chloro-n~1~-3,4-difluorophenyl benzamide, n-3,4-difluorophenyl 2-chlorophenyl carboxamide
C1=CC=C(C(=C1)C(=O)NC2=CC(=C(C=C2)F)F)Cl
267.66
267.66
97%
Videos
SDS