missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Chloro-N-(5-chloro-2-methylphenyl)benzamide, 97%

Brand:  Alfa Aesar™ H59316.03

Code : NEW

Additional Details : CAS Number : 199726-56-2 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15475588

  • 646.00 € / 1g

Estimated Shipment date
from Supplier 24-07-2019

Please to purchase this item. Need a web account? Register with us today!


Specifications

Chemical Identifiers

199726-56-2
280.148
AEGVAYXMYTXAKN-UHFFFAOYSA-N
295246
CC1=C(C=C(C=C1)Cl)NC(=O)C2=CC=CC=C2Cl
C14H11Cl2NO
MFCD00157769
2-chloro-n-5-chloro-2-methylphenyl benzamide, 2,5'-dichloro-2'-methylbenzanilide, 2-chloro-n-5-chloro-2-methyl-phenyl-benzamide, 2-chloro-n~1~-5-chloro-2-methylphenyl benzamide, n-5-chloro-2-methylphenyl 2-chlorophenyl carboxamide
2-chloro-N-(5-chloro-2-methylphenyl)benzamide

Specifications

2-Chloro-N-(5-chloro-2-methylphenyl)benzamide
199726-56-2
MFCD00157769
AEGVAYXMYTXAKN-UHFFFAOYSA-N
2-chloro-N-(5-chloro-2-methylphenyl)benzamide
295246
Solid
1g
C14H11Cl2NO
2-chloro-n-5-chloro-2-methylphenyl benzamide, 2,5'-dichloro-2'-methylbenzanilide, 2-chloro-n-5-chloro-2-methyl-phenyl-benzamide, 2-chloro-n~1~-5-chloro-2-methylphenyl benzamide, n-5-chloro-2-methylphenyl 2-chlorophenyl carboxamide
CC1=C(C=C(C=C1)Cl)NC(=O)C2=CC=CC=C2Cl
280.148
280.15
97%
Videos
SDS