missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Chloro-N-n-hexylbenzamide, 97%

Brand:  Alfa Aesar™ H59822.03

Code : NEW

Additional Details : CAS Number : 62797-98-2 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15476238

  • 646.00 € / 1g

Estimated Shipment date
from Supplier 24-07-2019

Please to purchase this item. Need a web account? Register with us today!


Specifications

Chemical Identifiers

62797-98-2
239.743
GZKUZAGDBBWXSH-UHFFFAOYSA-N
4456527
CCCCCCNC(=O)C1=CC=CC=C1Cl
C13H18ClNO
MFCD01212976
2-chloro-n-n-hexylbenzamide, benzamide, 2-chloro-n-hexyl
2-chloro-N-hexylbenzamide

Specifications

2-Chloro-N-n-hexylbenzamide
1g
C13H18ClNO
2-chloro-n-n-hexylbenzamide, benzamide, 2-chloro-n-hexyl
CCCCCCNC(=O)C1=CC=CC=C1Cl
239.743
239.75
97%
White to Yellow
62797-98-2
MFCD01212976
GZKUZAGDBBWXSH-UHFFFAOYSA-N
2-chloro-N-hexylbenzamide
4456527
Solid
Videos
SDS