Learn More
2-Chlorobenzoic acid, 98%, ACROS Organics™
Brand: Acros Organics 159245000
Code : 29
Additional Details : CAS Number : 118-91-2 Weight : 0.55000kg
Packaging | Plastic bottle |
---|---|
Quantity | 500g |
Chemical Identifiers
118-91-2 | |
156.57 | |
IKCLCGXPQILATA-UHFFFAOYSA-N | |
8374 | |
2-chlorobenzoic acid |
C7H5ClO2 | |
MFCD00002412 | |
o-chlorobenzoic acid, benzoic acid, o-chloro, benzoic acid, 2-chloro, 2-cba, kyselina o-chlorbenzoova, chlorobenzoic acid, ocba, benzoic acid, chloro, 2-chloro-benzoic acid, chlorobenzoate | |
CHEBI:30793 | |
C1=CC=C(C(=C1)C(=O)O)Cl |
Specifications
2-Chlorobenzoic acid | |
Authentic | |
285.0°C | |
139.0°C to 143.0°C | |
98% | |
97.5 | |
97.5% min. (HPLC) | |
ClC6H4CO2H | |
09, 334 | |
o-chlorobenzoic acid, benzoic acid, o-chloro, benzoic acid, 2-chloro, 2-cba, kyselina o-chlorbenzoova, chlorobenzoic acid, ocba, benzoic acid, chloro, 2-chloro-benzoic acid, chlorobenzoate | |
IKCLCGXPQILATA-UHFFFAOYSA-N | |
2-chlorobenzoic acid | |
8374 | |
156.57 | |
98% |
173°C | |
Plastic bottle | |
White | |
500g | |
118-91-2 | |
100.0 | |
C7H5ClO2 | |
MFCD00002412 | |
15, 2125 | |
Solubility in water: soluble in hot water. Other solubilities: freely soluble in alcohol and ether | |
C1=CC=C(C(=C1)C(=O)O)Cl | |
156.57 | |
CHEBI:30793 | |
Powder |
Safety and Handling
- 2-Chlorobenzoic acid
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.