missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chlorobenzylmagnesium chloride, 0.50M in 2-MeTHF, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mL
Description
It is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as pharmaceutical intermediate.
Solubility
Reacts with water.
Notes
Store at 4°C. Air and moisture sensitive.
It is used as pharmaceutical intermediate.
Solubility
Reacts with water.
Notes
Store at 4°C. Air and moisture sensitive.
Specifications
CAS | 29874-00-8 |
Density | 0.96 |
Molecular Formula | C7H6Cl2Mg |
MDL Number | MFCD01319895 |
Quantity | 100 mL |
UN Number | UN2924 |
Sensitivity | Air and moisture sensitive |
Synonym | 2-chlorobenzylmagnesium chloride, magnesium, chloro 2-chlorophenyl methyl, o-chlorobenzylmagnesium chloride, wdnzkgyawfimjl-uhfffaoysa-m, 2-chlorobenzyl magnesium chloride, chloro 2-chlorophenyl methyl magnesium, 2-chlorobenzylmagnesium chloride, 0.50m in 2-methf, 2-chlorobenzylmagnesium chloride 0.25 m in diethyl ether |
Solubility Information | Reacts with water. |
InChI Key | WDNZKGYAWFIMJL-UHFFFAOYSA-M |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'