missing translation for 'onlineSavingsMsg'
Learn More

2-(Chloromethyl)pyridine hydrochloride, 98%, Thermo Scientific Chemicals

Catalog Number 11483913
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
6959-47-3
C6H7Cl2N
164.029
MFCD00012811
JPMRGPPMXHGKRO-UHFFFAOYSA-N
2-chloromethyl pyridine hydrochloride, 2-picolyl chloride hydrochloride, 2-chloromethylpyridine hydrochloride, picolyl chloride hydrochloride, 2-chloromethyl pyridine hcl, 2-pyridylmethylchloride hydrochloride, 2-chloromethyl pyridinium chloride, pyridine, 2-chloromethyl-, hydrochloride, unii-yni4417kx0
23392
CHEBI:76600
2-(chloromethyl)pyridine;hydrochloride
C1=CC=NC(=C1)CCl.Cl
This item is not returnable. View return policy
6959-47-3
C6H7Cl2N
164.029
MFCD00012811
JPMRGPPMXHGKRO-UHFFFAOYSA-N
2-chloromethyl pyridine hydrochloride, 2-picolyl chloride hydrochloride, 2-chloromethylpyridine hydrochloride, picolyl chloride hydrochloride, 2-chloromethyl pyridine hcl, 2-pyridylmethylchloride hydrochloride, 2-chloromethyl pyridinium chloride, pyridine, 2-chloromethyl-, hydrochloride, unii-yni4417kx0
23392
CHEBI:76600
2-(chloromethyl)pyridine;hydrochloride
C1=CC=NC(=C1)CCl.Cl

2-(Chloromethyl)pyridine hydrochloride was used as reagent in base catalyzed alkylation of p-tert-butylcalix[6]arene and p-tert-butylcalix[5]arene in DMF. It was used in the synthesis of Gd3+ diethylenetriaminepentaacetic acid bisamide complex, a Zn2+-sensitive magnetic resonance imaging contrast agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Chloromethyl)pyridine hydrochloride was used as reagent in base catalyzed alkylation of p-tert-butylcalix[6]arene and p-tert-butylcalix[5]arene in DMF. It was used in the synthesis of Gd3+ diethylenetriaminepentaacetic acid bisamide complex, a Zn2+-sensitive magnetic resonance imaging contrast agent.

Solubility
Soluble in water, acetone (10-50mg/ml), 95%ethanol (100mg/ml).

Notes
Hygroscopic store in inert gas Store away from strong oxidizing agents. keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY
Melting Point 120°C to 124°C
Quantity 5 g
UN Number UN3261
Beilstein 3689155
Sensitivity Hygroscopic
Solubility Information Soluble in water,acetone (10-50mg/ml),95%ethanol (100mg/ml).
Formula Weight 164.04
Percent Purity 98%
Chemical Name or Material 2- (Chloromethyl)pyridine hydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'