missing translation for 'onlineSavingsMsg'
Learn More

2-Cyano-2,2-dimethylacetic acid, 95%, Thermo Scientific™

Catalog Number p-7057255
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
22426-30-8
C5H7NO2
113.12
MFCD11100210
DZDVNAVPCCNJRK-UHFFFAOYSA-N
2-cyano-2,2-dimethylacetic acid, 2-cyano-2-methyl-propanoic acid, cyanomethylpropanoicacid, 2-cyanoisobutyric acid, dimethylcyanoacetic acid, propanoic acid, 2-cyano-2-methyl
11367091
2-cyano-2-methylpropanoic acid
CC(C)(C#N)C(O)=O
This item is not returnable. View return policy
22426-30-8
C5H7NO2
113.12
MFCD11100210
DZDVNAVPCCNJRK-UHFFFAOYSA-N
2-cyano-2,2-dimethylacetic acid, 2-cyano-2-methyl-propanoic acid, cyanomethylpropanoicacid, 2-cyanoisobutyric acid, dimethylcyanoacetic acid, propanoic acid, 2-cyano-2-methyl
11367091
2-cyano-2-methylpropanoic acid
CC(C)(C#N)C(O)=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 56°C to 61°C
Color White
Boiling Point 132°C to 135°C (12 mmHg)
Quantity 250 mg
UN Number UN2923
Sensitivity Moisture sensitive
Formula Weight 113.11
Percent Purity 95%
Physical Form Crystalline Solid
Chemical Name or Material 2-Cyano-2,2-dimethylacetic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'