missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Cyano-2,2-dimethylacetic acid, 95%

Brand:  Alfa Aesar™ H66734.03

Code : NEW

Additional Details : CAS Number : 22426-30-8 Weight : 0.00100kg Transport : UN number : 2923 Chem class : 8 Pack group : III

Quantity 1g

 View more versions of this product

Product Code. 15472809

  • 392.00 € / 1g


Estimated Shipment date
from Supplier 22-05-2019
Add to basket

Specifications

Chemical Identifiers

22426-30-8
113.116
DZDVNAVPCCNJRK-UHFFFAOYSA-N
11367091
CC(C)(C#N)C(=O)O
C5H7NO2
MFCD11100210
2-cyano-2,2-dimethylacetic acid, 2-cyano-2-methyl-propanoic acid, cyanomethylpropanoicacid, 2-cyanoisobutyric acid, dimethylcyanoacetic acid, propanoic acid, 2-cyano-2-methyl
2-cyano-2-methylpropanoic acid

Specifications

2-Cyano-2,2-dimethylacetic acid
132°C to 135°C (12mmHg)
56°C to 61°C
22426-30-8
MFCD11100210
2-cyano-2,2-dimethylacetic acid, 2-cyano-2-methyl-propanoic acid, cyanomethylpropanoicacid, 2-cyanoisobutyric acid, dimethylcyanoacetic acid, propanoic acid, 2-cyano-2-methyl
CC(C)(C#N)C(=O)O
113.116
113.11
95%
Moisture Sensitive
White
1g
C5H7NO2
UN2923
DZDVNAVPCCNJRK-UHFFFAOYSA-N
2-cyano-2-methylpropanoic acid
11367091
Crystalline Solid
Videos
SDS