missing translation for 'onlineSavingsMsg'
Learn More

2-Cyanobenzaldehyde, 98%, Thermo Scientific Chemicals

Catalog Number p-7030478
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
7468-67-9
C8H5NO
131.13
MFCD00017503
QVTPWONEVZJCCS-UHFFFAOYSA-N
2-cyanobenzaldehyde, o-cyanobenzaldehyde, benzonitrile, 2-formyl, cyanobenzaldehyde, 2-formylbenzenecarbonitrile, 2-cyano-benzaldehyde, 2-formyl-benzonitrile, pubchem7409, #, acmc-1blgc
101209
2-formylbenzonitrile
O=CC1=CC=CC=C1C#N
This item is not returnable. View return policy
7468-67-9
C8H5NO
131.13
MFCD00017503
QVTPWONEVZJCCS-UHFFFAOYSA-N
2-cyanobenzaldehyde, o-cyanobenzaldehyde, benzonitrile, 2-formyl, cyanobenzaldehyde, 2-formylbenzenecarbonitrile, 2-cyano-benzaldehyde, 2-formyl-benzonitrile, pubchem7409, #, acmc-1blgc
101209
2-formylbenzonitrile
O=CC1=CC=CC=C1C#N

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 102°C to 106°C
Odor Characteristic
Quantity 1 g
Sensitivity Air Sensitive
Formula Weight 131.13
Percent Purity 98%
Chemical Name or Material 2-Cyanobenzaldehyde

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'