missing translation for 'onlineSavingsMsg'
Learn More

2-(Dicyclohexylphosphino)-2'-isopropylbiphenyl, 98%, Thermo Scientific Chemicals

Catalog Number 10015493
183.34 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
251320-85-1
C27H37P
392.56
MFCD09744023
PQWWLQRSCJJOIS-UHFFFAOYSA-N
2-dicyclohexylphosphino-2'-isopropylbiphenyl, 2-dicyclohexylphosphino-2'-i-propylbiphenyl, dicyclohexyl 2'-propan-2-yl 1,1'-biphenyl-2-yl phosphane, dicyclohexyl 2'-isopropyl-1,1'-biphenyl-2-yl phosphane
23420463
dicyclohexyl-[2-(2-propan-2-ylphenyl)phenyl]phosphane
CC(C)C1=CC=CC=C1C2=CC=CC=C2P(C3CCCCC3)C4CCCCC4
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
251320-85-1
C27H37P
392.56
MFCD09744023
PQWWLQRSCJJOIS-UHFFFAOYSA-N
2-dicyclohexylphosphino-2'-isopropylbiphenyl, 2-dicyclohexylphosphino-2'-i-propylbiphenyl, dicyclohexyl 2'-propan-2-yl 1,1'-biphenyl-2-yl phosphane, dicyclohexyl 2'-isopropyl-1,1'-biphenyl-2-yl phosphane
23420463
dicyclohexyl-[2-(2-propan-2-ylphenyl)phenyl]phosphane
CC(C)C1=CC=CC=C1C2=CC=CC=C2P(C3CCCCC3)C4CCCCC4

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble
Formula Weight 392.56
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2-(Dicyclohexylphosphino)-2'-isopropylbiphenyl
Show More Show Less
compliance-icons
Product Identifier
  • 2-(Dicyclohexylphosphino)-2'-isopropylbiphenyl
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'