missing translation for 'onlineSavingsMsg'
Learn More

2-(Ethylamino)ethanol, 98%, Thermo Scientific Chemicals

Catalog Number 10002080
16.06 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
250 g
1 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
110-73-6
C4H11NO
89.14
MFCD00002841
MIJDSYMOBYNHOT-UHFFFAOYSA-N
2-ethylamino ethanol, ethanol, 2-ethylamino, ethyl ethanolamine, n-ethylethanolamine, 2-hydroxyethyl ethylamine, n-ethylmonoethanolamine, 2-n-monoethylaminoethanol, n-ethylaminoethanol, n-ethyl ethanolamine, 2-ethylamino ethan-1-ol
8072
2-(ethylamino)ethanol
CCNCCO
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
110-73-6
C4H11NO
89.14
MFCD00002841
MIJDSYMOBYNHOT-UHFFFAOYSA-N
2-ethylamino ethanol, ethanol, 2-ethylamino, ethyl ethanolamine, n-ethylethanolamine, 2-hydroxyethyl ethylamine, n-ethylmonoethanolamine, 2-n-monoethylaminoethanol, n-ethylaminoethanol, n-ethyl ethanolamine, 2-ethylamino ethan-1-ol
8072
2-(ethylamino)ethanol
CCNCCO

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point -6°C
Color Colorless to Yellow
Density 0.9100g/mL
Boiling Point 167°C
Flash Point 78°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C2H5NHCH2CH2OH
Refractive Index 1.4390 to 1.441
Quantity 250 g
Beilstein 04, 282
Specific Gravity 0.91
Solubility Information Solubility in water: miscible. Other solubilities: soluble in alcohol and ether
Formula Weight 89.14
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-(Ethylamino)ethanol, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'