missing translation for 'onlineSavingsMsg'
Learn More

2-Ethynylpyridine, 98+%, Thermo Scientific Chemicals

Catalog Number p-3778588
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1945-84-2
C7H5N
103.12
MFCD00041598
NHUBNHMFXQNNMV-UHFFFAOYSA-N
2-ethynyl pyridine, 2-pyridylacetylene, pyridine, 2-ethynyl, 2-ethynyl-pyridine, pyridine, ethynyl, pyridin-2-yl acetylene, ethynylpyridine, ethynyl pyridine, ortho-ethynylpyridine
137268
2-ethynylpyridine
C#CC1=CC=CC=N1
This item is not returnable. View return policy
1945-84-2
C7H5N
103.12
MFCD00041598
NHUBNHMFXQNNMV-UHFFFAOYSA-N
2-ethynyl pyridine, 2-pyridylacetylene, pyridine, 2-ethynyl, 2-ethynyl-pyridine, pyridine, ethynyl, pyridin-2-yl acetylene, ethynylpyridine, ethynyl pyridine, ortho-ethynylpyridine
137268
2-ethynylpyridine
C#CC1=CC=CC=N1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.0
CAS Max % 100.0
Color Brown
Density 1.0200g/mL
Boiling Point 77.0°C to 78.0°C (14.0 mmHg)
Flash Point 76°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Refractive Index 1.5570 to 1.5610
Quantity 1 g
Specific Gravity 1.02
Solubility Information Solubility in water: soluble in water
Formula Weight 103.12
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material 2-Ethynylpyridine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'