missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Fluoro-3-(4-methylbenzylcarbamoyl)benzeneboronic acid, 97%

Brand:  Alfa Aesar™ H56951.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15472996

  • 671.00 € / 1g

Estimated Shipment date
from Supplier 18-07-2019

Please to purchase this item. Need a web account? Register with us today!


Specifications

Chemical Identifiers

1449145-04-3
287.10
OKJZHLHCULRTID-UHFFFAOYSA-N
73996259
C15H15BFNO3
MFCD22682901
2-fluoro-3-4-methylbenzylcarbamoyl benzeneboronic acid

Specifications

2-Fluoro-3-(4-methylbenzylcarbamoyl)benzeneboronic acid
1g
C15H15BFNO3
2-fluoro-3-4-methylbenzylcarbamoyl benzeneboronic acid
287.10
287.1
97%
White
1449145-04-3
MFCD22682901
OKJZHLHCULRTID-UHFFFAOYSA-N
73996259
Solid
Videos
SDS