missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Fluoro-4-nitrobenzoic acid, 95+%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
Specifications
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Formula Weight | 185.11 |
Percent Purity | 95% |
Chemical Name or Material | 2-Fluoro-4-nitrobenzoic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'