missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-nitrobenzoic acid, 95+%, Maybridge

Brand:  Maybridge TL00808DA

Code : 31

Additional Details : CAS Number : 403-24-7 Weight : 0.00100kg

Packaging Amber Glass Bottle
Quantity 1g

 View more versions of this product

Product Code. 10425532

  • 28.00 € / 1g


Estimated Shipment
21-09-2018
Add to basket

Specifications

Chemical Identifiers

403-24-7
185.11
302679
C7H4FNO4
MMWFMFZFCKADEL-UHFFFAOYSA-N

Specifications

2-Fluoro-4-nitrobenzoic acid
403-24-7
95%
C7H4FNO4
C1=CC(=C(C=C1[N+](=O)[O-])F)C(=O)O
302679
1g
Amber Glass Bottle
95%
MMWFMFZFCKADEL-UHFFFAOYSA-N
185.11
185.11
Videos
SDS