missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-5-methylpyridine, 97%, ACROS Organics™

Brand:  Acros Organics 443130010

Code : 29

Additional Details : CAS Number : 2369-19-9 Weight : 0.05100kg Transport : UN number : 1993 Chem class : 3 Pack group : III


Quantity 1g

 View more versions of this product

Product Code. 11956451

  • 42.70 € / 1g

Estimated Shipment: 06-07-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

2369-19-9
111.12
2-fluoro-5-picoline, 6-fluoro-3-picoline, 2-fluoro-5-methypyridine, 2-fluoro-5-methyl-pyridine, fluoromethylpyridine3, 2-fluoro-5-methyl pyridine, pyridine, 2-fluoro-5-methyl, pubchem4059, acmc-1cdos, 5-methyl-2-fluoropyridine
2-fluoro-5-methylpyridine
C6H6FN
AOSOZARHUJMBLZ-UHFFFAOYSA-N
2774635
CC1=CN=C(C=C1)F

Specifications

2-Fluoro-5-methylpyridine
Authentic
154.0°C to 155.0°C
1g
96.0
96% min. (GC)
2-fluoro-5-picoline, 6-fluoro-3-picoline, 2-fluoro-5-methypyridine, 2-fluoro-5-methyl-pyridine, fluoromethylpyridine3, 2-fluoro-5-methyl pyridine, pyridine, 2-fluoro-5-methyl, pubchem4059, acmc-1cdos, 5-methyl-2-fluoropyridine
CC1=CN=C(C=C1)F
111.12
111.12
97%
48°C
Glass Bottle
Colorless to Yellow
2369-19-9
100.0
C6H6FN
AOSOZARHUJMBLZ-UHFFFAOYSA-N
2-fluoro-5-methylpyridine
2774635
Liquid
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 2-Fluoro-5-methylpyridine

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS Documents
Certificates