missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-6-(trifluoromethyl)benzyl bromide, 95%, Thermo Scientific™

Catalog Number p-4812804
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
239087-08-2
C8H5BrF4
257.03
RINUERVPFANASB-UHFFFAOYSA-N
2-fluoro-6-trifluoromethyl benzyl bromide, 2-bromomethyl-1-fluoro-3-trifluoromethyl benzene, 2-fluoro-6-trifluoromethylbenzyl bromide, 2-fluoro-6-trifluoromethylbenzylbromide, 2-fluoro-6-trifluoromethyl benzylbromide, 2-bromomethyl-3-fluorobenzotrifluoride, alpha-bromo-2-fluoro-6-trifluoromethyl toluene, benzene, 2-bromomethyl-1-fluoro-3-trifluoromethyl, pubchem4932, acmc-1cg83
2737573
2-(bromomethyl)-1-fluoro-3-(trifluoromethyl)benzene
C1=CC(=C(C(=C1)F)CBr)C(F)(F)F
This item is not returnable. View return policy
239087-08-2
C8H5BrF4
257.03
RINUERVPFANASB-UHFFFAOYSA-N
2-fluoro-6-trifluoromethyl benzyl bromide, 2-bromomethyl-1-fluoro-3-trifluoromethyl benzene, 2-fluoro-6-trifluoromethylbenzyl bromide, 2-fluoro-6-trifluoromethylbenzylbromide, 2-fluoro-6-trifluoromethyl benzylbromide, 2-bromomethyl-3-fluorobenzotrifluoride, alpha-bromo-2-fluoro-6-trifluoromethyl toluene, benzene, 2-bromomethyl-1-fluoro-3-trifluoromethyl, pubchem4932, acmc-1cg83
2737573
2-(bromomethyl)-1-fluoro-3-(trifluoromethyl)benzene
C1=CC(=C(C(=C1)F)CBr)C(F)(F)F
Melting Point 42°C to 44°C
Flash Point 107°C
Assay Percent Range 94% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 257.03
Percent Purity 95%
Chemical Name or Material 2-Fluoro-6-(trifluoromethyl)benzyl bromide
compliance-icons
Product Identifier
  • 2-Fluoro-6-(trifluoromethyl)benzyl bromide
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'