missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-6-(trifluoromethyl)benzyl bromide, 95%, ACROS Organics™

Brand:  Acros Organics 442160050

Code : 29

Additional Details : CAS Number : 239087-08-2 Weight : 0.05500kg Transport : UN number : 3261 Chem class : 8 Pack group : III


Quantity 5g

 View more versions of this product

Product Code. 11974541

  • 115.00 € / 5g

In stock, ready to ship: 3

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

239087-08-2
257.03
2-fluoro-6-trifluoromethyl benzyl bromide, 2-bromomethyl-1-fluoro-3-trifluoromethyl benzene, 2-fluoro-6-trifluoromethylbenzyl bromide, 2-fluoro-6-trifluoromethylbenzylbromide, 2-fluoro-6-trifluoromethyl benzylbromide, 2-bromomethyl-3-fluorobenzotrifluoride, alpha-bromo-2-fluoro-6-trifluoromethyl toluene, benzene, 2-bromomethyl-1-fluoro-3-trifluoromethyl, pubchem4932, acmc-1cg83
2-(bromomethyl)-1-fluoro-3-(trifluoromethyl)benzene
C8H5BrF4
RINUERVPFANASB-UHFFFAOYSA-N
2737573
C1=CC(=C(C(=C1)F)CBr)C(F)(F)F

Specifications

2-Fluoro-6-(trifluoromethyl)benzyl bromide
42°C to 44°C
239087-08-2
C8H5BrF4
RINUERVPFANASB-UHFFFAOYSA-N
2-(bromomethyl)-1-fluoro-3-(trifluoromethyl)benzene
2737573
95%
107°C
5g
94% min. (GC)
2-fluoro-6-trifluoromethyl benzyl bromide, 2-bromomethyl-1-fluoro-3-trifluoromethyl benzene, 2-fluoro-6-trifluoromethylbenzyl bromide, 2-fluoro-6-trifluoromethylbenzylbromide, 2-fluoro-6-trifluoromethyl benzylbromide, 2-bromomethyl-3-fluorobenzotrifluoride, alpha-bromo-2-fluoro-6-trifluoromethyl toluene, benzene, 2-bromomethyl-1-fluoro-3-trifluoromethyl, pubchem4932, acmc-1cg83
C1=CC(=C(C(=C1)F)CBr)C(F)(F)F
257.03
257.03
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 2-Fluoro-6-(trifluoromethyl)benzyl bromide

Signal Word
  • Danger

Hazard Category
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • EUH029-Contact with water liberates toxic gas.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

SDS Documents