missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Fluoro-N-(2,5-difluorophenyl)benzamide, 97%

Brand:  Alfa Aesar™ H59526.03

Code : NEW

Additional Details : CAS Number : 903269-74-9 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15415878

  • 448.12 € / 1g


Estimated Shipment date
from Supplier 30-07-2018
Add to basket

Specifications

Chemical Identifiers

2-Fluoro-N-(2,5-difluorophenyl)benzamide
903269-74-9
MFCD04566569

Specifications

2-Fluoro-N-(2,5-difluorophenyl)benzamide
1g
Solid
C13H8F3NO
PTYNUUHSBNFFRC-UHFFFAOYSA-N
251.208
251.21
903269-74-9
97%
MFCD04566569
C1=CC=C(C(=C1)C(=O)NC2=C(C=CC(=C2)F)F)F
2711386
Videos
SDS