missing translation for 'onlineSavingsMsg'
Learn More

(2-Fluorophenyl)methanesulfonyl chloride, ≥95%, Thermo Scientific™

Catalog Number p-3175190
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
24974-71-8
C7H6ClFO2S
208.63
MFCD04117455
CJDWSUDXKHLMCY-UHFFFAOYSA-N
2-fluorophenyl methanesulfonyl chloride, 2-fluoro-phenyl-methanesulfonyl chloride, 2-fluorobenzylsulfonyl chloride, 2-fluorophenyl methanesulphonyl chloride, chloro 2-fluorophenyl methyl sulfone, alpha-chlorosulphonyl-2-fluorotoluene, benzenemethanesulfonyl chloride, 2-fluoro, 2-fluorophenyl-methanesulfonyl chloride, pubchem10055
573052
(2-fluorophenyl)methanesulfonyl chloride
FC1=CC=CC=C1CS(Cl)(=O)=O
This item is not returnable. View return policy
24974-71-8
C7H6ClFO2S
208.63
MFCD04117455
CJDWSUDXKHLMCY-UHFFFAOYSA-N
2-fluorophenyl methanesulfonyl chloride, 2-fluoro-phenyl-methanesulfonyl chloride, 2-fluorobenzylsulfonyl chloride, 2-fluorophenyl methanesulphonyl chloride, chloro 2-fluorophenyl methyl sulfone, alpha-chlorosulphonyl-2-fluorotoluene, benzenemethanesulfonyl chloride, 2-fluoro, 2-fluorophenyl-methanesulfonyl chloride, pubchem10055
573052
(2-fluorophenyl)methanesulfonyl chloride
FC1=CC=CC=C1CS(Cl)(=O)=O
CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 208.64
Percent Purity 95%
Chemical Name or Material (2-Fluorophenyl)methanesulfonyl chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'