Learn More
2-Hexanone, 98%, ACROS Organics™
2-Hexanone, 98%, C6H12O, CAS Number-591-78-6, 2-hexanone, 2-oxohexane, butyl methyl ketone, hexanone, hexanone-2, ketone, butyl methyl, methyl butyl ketone, methyl n-butyl ketone, n-butyl methyl ketone, propylacetone, 500mL, 01, 689, 127 deg.C, 100.0, 97.5, Colorless to Yellow, 0.8100g/mL, 209-731-1
Brand: Acros Organics 146885000
Code : 29
Additional Details : CAS Number : 591-78-6 Weight : 0.45500kg Transport : UN number : 1224 Chem class : 3 Pack group : III
Packaging | Glass bottle |
---|---|
Quantity | 500mL |
Description
2-Hexanone, 98%, C6H12O, CAS Number-591-78-6, 2-hexanone, 2-oxohexane, butyl methyl ketone, hexanone, hexanone-2, ketone, butyl methyl, methyl butyl ketone, methyl n-butyl ketone, n-butyl methyl ketone, propylacetone, 500mL, 01, 689, 127 deg.C, 100.0, 97.5, Colorless to Yellow, 0.8100g/mL, 209-731-1
Chemical Identifiers
591-78-6 | |
100.16 | |
QQZOPKMRPOGIEB-UHFFFAOYSA-N | |
11583 | |
CCCCC(=O)C |
C6H12O | |
MFCD00009482 | |
2-hexanone, hexanone, 2-oxohexane, propylacetone, n-butyl methyl ketone, butyl methyl ketone, methyl butyl ketone, methyl n-butyl ketone, hexanone-2, ketone, butyl methyl | |
hexan-2-one |
Specifications
2-Hexanone | |
23°C | |
Glass bottle | |
0.81 | |
Colorless to Yellow | |
500mL | |
591-78-6 | |
100.0 | |
C6H12O | |
MFCD00009482 | |
15, 6107 | |
Solubility in water: 20g/l (20°C). Other solubilities: soluble in alcohol and ether | |
CCCCC(=O)C | |
100.16 | |
100.16 | |
98% |
0.8100g/mL | |
Authentic | |
1.3995 to 1.4015 | |
127°C | |
-57°C | |
98% | |
97.5 | |
98% | |
CH3(CH2)3COCH3 | |
01, 689 | |
2-hexanone, hexanone, 2-oxohexane, propylacetone, n-butyl methyl ketone, butyl methyl ketone, methyl butyl ketone, methyl n-butyl ketone, hexanone-2, ketone, butyl methyl | |
QQZOPKMRPOGIEB-UHFFFAOYSA-N | |
hexan-2-one | |
11583 | |
Liquid |
Safety and Handling
- 2-Hexanone
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 3
- Reproductive toxicity Category 2
- Specific target organ toxicity Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H372-Causes damage to organs.
- H361f-Suspected of damaging fertility.
- H336-May cause drowsiness or dizziness.
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.