Learn More
2-Hydroxy-4-methoxybenzophenone, 98+%, Thermo Scientific Chemicals
Description
2-Hydroxy-4-methoxybenzophenone is used as a broad-band filter in sunscreens cosmetic products in order to protect the skin from the deleterious effects of the sun. It acts as a reagent in the gravimetric determination of copper(II). Further, it is used in the preparation of poly[(2-hydroxy-4-methoxybenzophenone) propylene], which is a polymeric ligand useful for the synthesis of metal/ligand polychelates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2-Hydroxy-4-methoxybenzophenone is used as a broad-band filter in sunscreens cosmetic products in order to protect the skin from the deleterious effects of the sun. It acts as a reagent in the gravimetric determination of copper(II). Further, it is used in the preparation of poly[(2-hydroxy-4-methoxybenzophenone) propylene], which is a polymeric ligand useful for the synthesis of metal/ligand polychelates.
Solubility
Insoluble in water.
Notes
Store in a cool place. Incompatible with bases, acid chlorides, acid anhydrides and oxidizing agents.
Specifications
Melting Point | 62°C to 65°C |
Density | 1.3 |
Boiling Point | ∼150°C (5mmHg) |
Flash Point | 216°C (420°F) |
Quantity | 50 g |
Beilstein | 1913145 |
Merck Index | 14,6954 |
Solubility Information | Insoluble in water. |
Formula Weight | 228.25 |
Percent Purity | ≥98% |
Show More |
RUO – Research Use Only