missing translation for 'onlineSavingsMsg'
Learn More

2-Iminothiolane hydrochloride, 98%, Thermo Scientific Chemicals

Catalog Number p-4812814
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
4781-83-3
C4H7NS·ClH
137.63
ATGUDZODTABURZ-UHFFFAOYSA-N
2-iminothiolane hydrochloride, 2-iminothiolane hcl, thiolan-2-imine hydrochloride, dihydrothiophen-2 3h-imine hydrochloride, 2 3h-thiophenimine, dihydro-, hydrochloride, traut's reagent, c4h7ns.clh, traut eo 1/4 a, 2-thiolanimine hydrochloride, 2 3h-thiophenimine, hydrochloride
13166855
thiolan-2-imine;hydrochloride
C1CC(=N)SC1.Cl
This item is not returnable. View return policy
4781-83-3
C4H7NS·ClH
137.63
ATGUDZODTABURZ-UHFFFAOYSA-N
2-iminothiolane hydrochloride, 2-iminothiolane hcl, thiolan-2-imine hydrochloride, dihydrothiophen-2 3h-imine hydrochloride, 2 3h-thiophenimine, dihydro-, hydrochloride, traut's reagent, c4h7ns.clh, traut eo 1/4 a, 2-thiolanimine hydrochloride, 2 3h-thiophenimine, hydrochloride
13166855
thiolan-2-imine;hydrochloride
C1CC(=N)SC1.Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Assay Percent Range 97.5% min. (Argentometry)
Packaging Glass Bottle
Quantity 250 mg
Formula Weight 137.63
Percent Purity 98%
Chemical Name or Material 2-Iminothiolane Hydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'