missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Iodoaniline, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
50 g
100 g
Description
2-Iodoaniline is used as an intermediate in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Iodoaniline is used as an intermediate in organic synthesis.
Solubility
Insoluble in water.
Notes
It is sensitive to light. Incompatible with oxidizing agents.
2-Iodoaniline is used as an intermediate in organic synthesis.
Solubility
Insoluble in water.
Notes
It is sensitive to light. Incompatible with oxidizing agents.
Specifications
Melting Point | 53°C to 57°C |
Flash Point | >110°C (230°F) |
Quantity | 5 g |
Beilstein | 2204899 |
Sensitivity | Light sensitive |
Solubility Information | Insoluble in water. |
Formula Weight | 219.03 |
Percent Purity | ≥98% |
Chemical Name or Material | 2-Iodoaniline |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'