Learn More
2-Iodobenzotrifluoride, 99%, ACROS Organics™
Brand: Acros Organics 241980050
Code : 29
Additional Details : CAS Number : 444-29-1 Weight : 0.05945kg
Packaging | Glass bottle |
---|---|
Quantity | 5mL |
Chemical Identifiers
444-29-1 | |
272.01 | |
IGZGUYVVBABKOY-UHFFFAOYSA-N | |
67957 | |
C1=CC=C(C(=C1)C(F)(F)F)I |
C7H4F3I | |
MFCD00001041 | |
2-iodobenzotrifluoride, 1-iodo-2-trifluoromethyl benzene, o-iodobenzotrifluoride, benzene, 1-iodo-2-trifluoromethyl, 2-iodobenztrifluoride, o-trifluoromethyliodobenzene, 2-iodo-alpha,alpha,alpha-trifluorotoluene, iodobenzotrifluoride, 2-iodo-a,a,a-trifluorotoluene, 2-iodobenzotrifloride | |
1-iodo-2-(trifluoromethyl)benzene |
Specifications
2-Iodobenzotrifluoride | |
81°C | |
Glass bottle | |
1.89 | |
Yellow | |
444-29-1 | |
100.0 | |
C7H4F3I | |
MFCD00001041 | |
2-iodobenzotrifluoride, 1-iodo-2-trifluoromethyl benzene, o-iodobenzotrifluoride, benzene, 1-iodo-2-trifluoromethyl, 2-iodobenztrifluoride, o-trifluoromethyliodobenzene, 2-iodo-alpha,alpha,alpha-trifluorotoluene, iodobenzotrifluoride, 2-iodo-a,a,a-trifluorotoluene, 2-iodobenzotrifloride | |
IGZGUYVVBABKOY-UHFFFAOYSA-N | |
1-iodo-2-(trifluoromethyl)benzene | |
67957 | |
Liquid |
1.8900g/mL | |
Authentic | |
1.5290 to 1.5310 | |
197.0°C to 198.0°C | |
5mL | |
98.5 | |
98.5% min. (GC) | |
CF3(C6H4)I | |
05, III, 725 | |
Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents | |
C1=CC=C(C(=C1)C(F)(F)F)I | |
272.01 | |
272.01 | |
99% |
Safety and Handling
- 2-Iodobenzotrifluoride
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.