missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-3-pyridinecarboxaldehyde, 96%, ACROS Organics™

Brand:  Acros Organics 439780010

Code : 29

Additional Details : CAS Number : 71255-09-9 Weight : 0.05100kg


Packaging Glass bottle
Quantity 1g

 View more versions of this product

Product Code. 11956391

  • 60.78 € / 1g

Estimated Shipment: 18-08-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

71255-09-9
137.14
2-methoxynicotinaldehyde, 2-methoxy-3-pyridinecarboxaldehyde, 2-methoxy-pyridine-3-carbaldehyde, 2-methoxypyridine-3-carboxaldehyde, 3-pyridinecarboxaldehyde, 2-methoxy, 2-methoxy-3-pyridine carboxaldehyde, 3-pyridinecarboxaldehyde,2-methoxy, 2-methoxy-3-pyridinecarbaldehyde, pubchem15124, 2-methoxy-3-formyl-pyridine
2-methoxypyridine-3-carbaldehyde
C7H7NO2
PIFFMIDNNWOQLK-UHFFFAOYSA-N
12533391
COC1=C(C=CC=N1)C=O

Specifications

2-Methoxy-3-pyridinecarboxaldehyde
Authentic
200°C to 201°C
25°C
71255-09-9
100.0
C7H7NO2
PIFFMIDNNWOQLK-UHFFFAOYSA-N
2-methoxypyridine-3-carbaldehyde
12533391
Liquid or Low Melting Solid
96°C
Glass bottle
Colorless to Yellow
1g
95.0
95% min. (GC)
2-methoxynicotinaldehyde, 2-methoxy-3-pyridinecarboxaldehyde, 2-methoxy-pyridine-3-carbaldehyde, 2-methoxypyridine-3-carboxaldehyde, 3-pyridinecarboxaldehyde, 2-methoxy, 2-methoxy-3-pyridine carboxaldehyde, 3-pyridinecarboxaldehyde,2-methoxy, 2-methoxy-3-pyridinecarbaldehyde, pubchem15124, 2-methoxy-3-formyl-pyridine
COC1=C(C=CC=N1)C=O
137.14
137.14
96%
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 2-Methoxy-3-pyridinecarboxaldehyde

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

SDS Documents
Certificates