missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1,3-benzothiazole-6-sulfonyl chloride, 95%, Maybridge

Brand:  Maybridge CC54703DE

Code : 31

Additional Details : CAS Number : 21431-13-0 Weight : 0.05500kg Transport : UN number : 3261 Chem class : 8 Pack group : III

Packaging Amber Glass Bottle
Quantity 5g

 View more versions of this product

Product Code. 10001154

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 01 885 5854  for assistance.


Specifications

Chemical Identifiers

21431-13-0
247.711
2-methyl-benzothiazole-6-sulfonyl chloride, 2-methylbenzo d thiazole-6-sulfonyl chloride, chloro 2-methylbenzothiazol-6-yl sulfone, 2-methylbenzothiazole-6-sulfonyl chloride, 6-benzothiazolesulfonyl chloride, 2-methyl, 1,3-benzothiazole-6-sulfonoyl chloride, 2-methyl
2-methyl-1,3-benzothiazole-6-sulfonyl chloride
C8H6ClNO2S2
PMMBRDLKYRRHMI-UHFFFAOYSA-N
3583019
CC1=NC2=C(S1)C=C(C=C2)S(=O)(=O)Cl

Specifications

2-Methyl-1, 3-benzothiazole-6-sulfonyl chloride
5g
95%
2-methyl-benzothiazole-6-sulfonyl chloride, 2-methylbenzo d thiazole-6-sulfonyl chloride, chloro 2-methylbenzothiazol-6-yl sulfone, 2-methylbenzothiazole-6-sulfonyl chloride, 6-benzothiazolesulfonyl chloride, 2-methyl, 1,3-benzothiazole-6-sulfonoyl chloride, 2-methyl
CC1=NC2=C(S1)C=C(C=C2)S(=O)(=O)Cl
247.711
247.73
Amber Glass Bottle
21431-13-0
C8H6ClNO2S2
PMMBRDLKYRRHMI-UHFFFAOYSA-N
2-methyl-1,3-benzothiazole-6-sulfonyl chloride
3583019
95%
Videos
SDS