missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1,5-diphenyl-1H-pyrrole-3-carboxylic acid, 97%, Thermo Scientific™

Click to view available options
Quantity:
1g
5g
10g
109812-64-8
C18H15NO2
277.323
MFCD01567220
DWIYTBRYOQDHTE-UHFFFAOYSA-N
2-methyl-1,5-diphenyl-1h-pyrrole-3-carboxylic acid, 1,5-diphenyl-2-methylpyrrole-3-carboxylic acid, 2-methyl-1,5-diphenyl-3-pyrrolecarboxylic acid, 1h-pyrrole-3-carboxylicacid, 2-methyl-1,5-diphenyl, maybridge1_008589, acmc-20mclm, 2-methyl-1,5-diphenyl-pyrrole-3-carboxylic acid
2735512
2-methyl-1,5-diphenylpyrrole-3-carboxylic acid
CC1=C(C=C(N1C2=CC=CC=C2)C3=CC=CC=C3)C(=O)O
109812-64-8
C18H15NO2
277.323
MFCD01567220
DWIYTBRYOQDHTE-UHFFFAOYSA-N
2-methyl-1,5-diphenyl-1h-pyrrole-3-carboxylic acid, 1,5-diphenyl-2-methylpyrrole-3-carboxylic acid, 2-methyl-1,5-diphenyl-3-pyrrolecarboxylic acid, 1h-pyrrole-3-carboxylicacid, 2-methyl-1,5-diphenyl, maybridge1_008589, acmc-20mclm, 2-methyl-1,5-diphenyl-pyrrole-3-carboxylic acid
2735512
2-methyl-1,5-diphenylpyrrole-3-carboxylic acid
CC1=C(C=C(N1C2=CC=CC=C2)C3=CC=CC=C3)C(=O)O
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1g
Formula Weight 277.32
Percent Purity 97%
Chemical Name or Material 2-Methyl-1, 5-diphenyl-1H-pyrrole-3-carboxylic acid