missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-3-nitroanisole, 99%, Thermo Scientific™

Catalog Number 11954711
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy
4837-88-1
C8H9NO3
167.16
MFCD00007159
HQCZLEAGIOIIMC-UHFFFAOYSA-N
2-methyl-3-nitroanisole, 2-methoxy-6-nitrotoluene, benzene, 1-methoxy-2-methyl-3-nitro, 1-methoxy-2-methyl-3-nitro-benzene, anisole, 2-methyl-3-nitro, 6-methoxy-2-nitrotoluene, 1-meo-2-me-3-no2-benzene, 3-nitro-2-methyl anisole, 3-methoxy-2-methyl-1-nitrobenzene, 2-methyl-3-nitroanisole 2-methoxy-6-nitrotoluene
78554
1-methoxy-2-methyl-3-nitrobenzene
COC1=CC=CC(=C1C)[N+]([O-])=O
This item is not returnable. View return policy
4837-88-1
C8H9NO3
167.16
MFCD00007159
HQCZLEAGIOIIMC-UHFFFAOYSA-N
2-methyl-3-nitroanisole, 2-methoxy-6-nitrotoluene, benzene, 1-methoxy-2-methyl-3-nitro, 1-methoxy-2-methyl-3-nitro-benzene, anisole, 2-methyl-3-nitro, 6-methoxy-2-nitrotoluene, 1-meo-2-me-3-no2-benzene, 3-nitro-2-methyl anisole, 3-methoxy-2-methyl-1-nitrobenzene, 2-methyl-3-nitroanisole 2-methoxy-6-nitrotoluene
78554
1-methoxy-2-methyl-3-nitrobenzene
COC1=CC=CC(=C1C)[N+]([O-])=O
CAS Min % 98.5
CAS Max % 100.0
Melting Point 54.0°C to 55.0°C
Color Brown to Yellow
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass Bottle
Linear Formula CH3C6H3(NO2)OCH3
Quantity 1 g
Beilstein 06,I,178
Formula Weight 167.16
Percent Purity 99%
Physical Form Crystalline Powder or Powder
Chemical Name or Material 2-Methyl-3-nitroanisole
Show More Show Less
compliance-icons
Product Identifier
  • 2-Methyl-3-nitroanisole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'