missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-3-(trifluoromethyl)aniline, 97%, Thermo Scientific™

Catalog Number 11975211
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
54396-44-0
C8H8F3N
175.16
TWLDBACVSHADLI-UHFFFAOYSA-N
2-methyl-3-trifluoromethyl aniline, 3-amino-2-methylbenzotrifluoride, 3-trifluoromethyl-o-toluidine, 3-amino-2-methyl benzotrifluoride, 2-methyl-3-trifluoromethyl-phenylamine, 2-methyl-3 trifluoromethyl aniline, 2-methyl-3-trifluoromethyl benzenamine, benzenamine, 2-methyl-3-trifluoromethyl
2735932
2-methyl-3-(trifluoromethyl)aniline
CC1=C(C=CC=C1N)C(F)(F)F
This item is not returnable. View return policy
54396-44-0
C8H8F3N
175.16
TWLDBACVSHADLI-UHFFFAOYSA-N
2-methyl-3-trifluoromethyl aniline, 3-amino-2-methylbenzotrifluoride, 3-trifluoromethyl-o-toluidine, 3-amino-2-methyl benzotrifluoride, 2-methyl-3-trifluoromethyl-phenylamine, 2-methyl-3 trifluoromethyl aniline, 2-methyl-3-trifluoromethyl benzenamine, benzenamine, 2-methyl-3-trifluoromethyl
2735932
2-methyl-3-(trifluoromethyl)aniline
CC1=C(C=CC=C1N)C(F)(F)F
CAS Min % 96.0
CAS Max % 100.0
Melting Point 39.0°C to 41.0°C
Color White
Boiling Point 108.0°C to 110.0°C
Flash Point 96°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Refractive Index 1.4825
Quantity 1 g
Formula Weight 175.16
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2-Methyl-3-(trifluoromethyl)aniline
Show More Show Less
compliance-icons
Product Identifier
  • 2-Methyl-3-(trifluoromethyl)aniline
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'