missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Methylthio-4(3H)-pyrimidinone, cont. ca 12% 6-hydroxy-2-methylthio-4(3H)-pyrimidinone

Brand:  Alfa Aesar™ H63154.06

Code : NEW

Additional Details : CAS Number : 5751-20-2 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15415595

  • 53.80 € / 5g


Estimated Shipment date
from Supplier 21-11-2018
Add to basket

Specifications

Chemical Identifiers

5751-20-2
142.176
UYHSQVMHSFXUOA-UHFFFAOYSA-N
79823
CSC1=NC=CC(=O)N1
C5H6N2OS
MFCD00047373
2-methylthio pyrimidin-4-ol, 2-methylthiopyrimidin-4-ol, 2-methylthio-4-pyrimidinol, 2-methylthio pyrimidin-4 3h-one, 2-methylthio-4-pyrimidone, 2-methylthiouracil, 2-methylsulfanyl pyrimidin-4-ol, 2-methylthio-4-pyrimidinol, 2-methylthio-4-hydroxypyrimidine, 4 1h-pyrimidinone, 2-methylthio
2-methylsulfanyl-1H-pyrimidin-6-one

Specifications

2-Methylthio-4(3H)-pyrimidinone
5g
5751-20-2
MFCD00047373
UYHSQVMHSFXUOA-UHFFFAOYSA-N
2-methylsulfanyl-1H-pyrimidin-6-one
79823
Solid
White
cont. ≈12% 6-hydroxy-2-methylthio-4(3H)-pyrimidinone
C5H6N2OS
2-methylthio pyrimidin-4-ol, 2-methylthiopyrimidin-4-ol, 2-methylthio-4-pyrimidinol, 2-methylthio pyrimidin-4 3h-one, 2-methylthio-4-pyrimidone, 2-methylthiouracil, 2-methylsulfanyl pyrimidin-4-ol, 2-methylthio-4-pyrimidinol, 2-methylthio-4-hydroxypyrimidine, 4 1h-pyrimidinone, 2-methylthio
CSC1=NC=CC(=O)N1
142.176
142.18
Videos
SDS