missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-N-Butoxyethanol, Extra Pure, SLR, Fisher Chemical™
Click to view available options
Quantity:
500 mL
2.5 L
25 L
Packaging:
Metal can
Plastic Coated Glass bottle
Description
2-N-Butoxyethanol, Extra Pure, SLR
Specifications
Melting Point | -70°C |
Color | Yellow |
Boiling Point | 171°C |
Packaging | Metal can |
Quantity | 25 L |
Formula Weight | 118.18g/mol |
Vapor Pressure | 0.8 hPa at 20°C |
Physical Form | Liquid |
Safety and Handling
Product Identifier
- 2-Butoxyethanol
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'