missing translation for 'onlineSavingsMsg'
Learn More

2-n-Pentylpyrazine, 97%, Thermo Scientific™

Catalog Number p-7058271
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
6303-75-9
C9H14N2
MFCD00014614
n-Amylpyrazine; 1-Pyrazinylpentane
This item is not returnable. View return policy
6303-75-9
C9H14N2
MFCD00014614
n-Amylpyrazine; 1-Pyrazinylpentane

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Density 0.95
Boiling Point 218°C
Flash Point 81°C (178°F)
Refractive Index 1.49
Quantity 1 g
Beilstein 508952
Formula Weight 150.23
Percent Purity 97%
Chemical Name or Material 2-n-Pentylpyrazine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'